2,6-Diisopropylaniline
Catalog # ABCRAB141935.0500
Supplier: abcr
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mL
Specifications
About this item
:
:177.29 g/mol
Boiling Pt:257 °C (1013 hPa)
Melting Pt:–45 °C
Density:0,94 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:246-305-4
CAS Number:24544-04-5
Formula:C₁₂H₁₉N
MDL Number:MFCD00008887