
1,1-Bis(methylthio)-2-nitroethylene, Sigma-Aldrich®
Catalog # SIAL279706-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:165.24 g/mol
Melting Pt:122…125 °C
Storage Temperature:Refrigerator
UN:2811
ADR:6.1,II
:M-Clarity™ quality level = MQ100
EINECS:237-108-4
:GHS07
CAS Number:13623-94-4
Formula:C₄H₇NO₂S₂
MDL Number:MFCD00010443