Order Entry
Czech Republic
ContactUsLinkComponent
Disodium hydrogen phosphate heptahydrate 98-100.0% (dried basis), crystals USP, J.T.Baker®
Disodium hydrogen phosphate heptahydrate 98-100.0% (dried basis), crystals USP, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Disodium hydrogen phosphate heptahydrate 98-100.0% (dried basis), crystals USP, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • GMP Manufactured Product:
    98.0 - 100.5%
  • Meets U.S.P Requirements:
    ≤16 ppm
  • CAUTION: For Manufacturing, processing or repackaging:
    ≤0.06%
  • Bulk Pharmaceutical Chemical:
    Passes Test
  • USP - Assay (Na₂HPO₄) (dried basis):
    98.0 - 100.5 %
  • USP - Arsenic (As):
    ≤16 ppm
  • USP - Chloride (Cl):
    ≤0.06 %
  • USP - Identification:
    Passes Test
  • USP - Insoluble Substances:
    ≤0.4 %
  • USP - Loss on Drying at 130°C:
    43.0 - 50.0 %
  • USP - Sulfate (SO₄):
    ≤0.2 %

Specification Test Results

Related Information

Frequently Bought Together