

Benzene-[D₆] (99.5% D)
Supplier: abcr
Certificates
About this item
CAS Number:1076-43-3
:
Formula:C₆D₆
MDL Number:MFCD00003010
:84.07 g/mol
Boiling Pt:79 °C (1013 hPa)
Melting Pt:6.7 °C
Density:0.95 g/cm³ (20 °C)
Flash Pt:–11 °C (closed cup)
Storage Temperature:Ambient
EINECS:214-061-8
UN:1114
ADR:3,II
:GHS02
:GHS08
Frequently Bought Together