Order Entry
Czech Republic
ContactUsLinkComponent
Boron standard solution, 1,000 mg/L B in water standard for AAS
Boron standard solution, 1,000 mg/L B in water standard for AAS
Catalog # REGEAAB-H
Supplier:  REAGECON
CAS Number:  
Boron standard solution, 1,000 mg/L B in water standard for AAS
Catalog # REGEAAB-H
Supplier:  REAGECON
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    H₂O
  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Element:
    Boron
  • Symbol:
    B

Specifications

About this item