Order Entry
Czech Republic
ContactUsLinkComponent
Arsenic standard solution, 1000 mg/L As in dil. nitric acid (from As₂O₃) TraceCERT® standard for ICP, Supelco®
Arsenic standard solution, 1000 mg/L As in dil. nitric acid (from As₂O₃) TraceCERT® standard for ICP, Supelco®
Catalog # SUPL01969-100ML-F
Supplier:  Merck
CAS Number:  
Arsenic standard solution, 1000 mg/L As in dil. nitric acid (from As₂O₃) TraceCERT® standard for ICP, Supelco®
Catalog # SUPL01969-100ML-F
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    2% HNO₃
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • M-Clarity quality level:
    MQ100
  • Element:
    Arsenic
  • Symbol:
    As

Specifications

About this item

Certifications: Traceable to NIST, tested in ISO 17034 /ISO17025 accredited laboratory.