Order Entry
Czech Republic
ContactUsLinkComponent
 
Arsenic standard solution, 1000 mg/L As in dil. nitric acid standard for ICP
  REGEPAS2A2
 :  REAGECON
 :  
Arsenic standard solution, 1000 mg/L As in dil. nitric acid standard for ICP
  REGEPAS2A2
 :  REAGECON
 :  

 

  • Concentration:
    1000 mg/L
  • Matrix:
    2-5% HNO₃
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Element:
    Arsenic
  • Symbol:
    As