Octamethyltrisiloxane, 1.0 cSt
ABCRAB112069.0100
: abcr
:
- Pack type:Glass bottle for solids
- Pk:100 g
:
:236.53 g/mol
:12,06844
:153 °C (1013 hPa)
:–82 °C
:0.82 g/cm³ (20 °C)
:29 °C (closed cup)
:203-497-4
:1993
:3,III
:GHS02
:107-51-7
:[(CH₃)₃SiO]₂Si(CH₃)₂
:MFCD00008264