Order Entry
Czech Republic
ContactUsLinkComponent
Polydimethylsiloxane, 5000 cSt
Polydimethylsiloxane, 5000 cSt
Catalog # ABCRAB112161.0100
Supplier:  abcr
CAS Number:  
Polydimethylsiloxane, 5000 cSt
Catalog # ABCRAB112161.0100
Supplier:  abcr
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
CAS Number:63148-62-9
 :9016-00-6
Formula:(CH₃)₃SiO(Si(CH₃)₂O)n
MDL Number:MFCD00084411