Polydimethylsiloxane, 10000 cSt
Catalog # ABCRAB112162.0100
Supplier: abcr
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
CAS Number:63148-62-9
:9016-00-6
Formula:(CH₃)₃SiO(Si(CH₃)₂O)n
MDL Number:MFCD00084411