0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
Potassium hydroxide
Potassium hydroxide 0.5 mol/l (0.5 N) in aqueous solution
Potassium hydroxide 0.5 mol/l (0.5 N) in aqueous solution
REGEKOH20501
:
REAGECON
:
Potassium hydroxide 0.5 mol/l (0.5 N) in aqueous solution
REGEKOH20501
:
REAGECON
:
Pack type:
Glass bottle
Pk:
1 L
Seq:
5037
:
:
56.11 g/mol
:
Ambient
:
215-181-3
:
1814
:
8,III
:
GHS05
:
1310-58-3
:
KOH
:
MFCD00003553