Lead standard solution, 1000 mg/L Pb in 0.5 M nitric acid standard for AAS
Catalog # REGEAAPBH
Supplier: REAGECON
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:0,5 M HNO₃
- Pack type:Glass bottle
- Pk:500 mL
- Element:Lead
- Symbol:Pb