Order Entry
Czech Republic
ContactUsLinkComponent
Ammonium standard solution, 1000 mg/L NH₄ in water standard for Ion Chromatography
Ammonium standard solution, 1000 mg/L NH₄ in water standard for Ion Chromatography
Catalog # 713-4073
Supplier:  WTW
CAS Number:  
Ammonium standard solution, 1000 mg/L NH₄ in water standard for Ion Chromatography
Catalog # 713-4073
Supplier:  WTW
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    H₂O
  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Colour:
    Colourless
  • Description:
    Ammonium standard solution
  • Environmentally Preferable:
  • Solubility:
    Soluble in water
  • Physical form:
    Liquid
  • pH range:
    0,5

Specifications

About this item