Order Entry
Czech Republic
ContactUsLinkComponent
Iron(II) sulphate heptahydrate ≥99% (by titrimetric analysis), granules, BAKER ANALYZED® ACS, J.T.Baker®
Irritant
Iron(II) sulphate heptahydrate ≥99% (by titrimetric analysis), granules, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Irritant
Iron(II) sulphate heptahydrate ≥99% (by titrimetric analysis), granules, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    99.0 - 104.5 %
  • Assay (FeSO₄·7H₂O) (by KMnO₄ titrn):
    99.0 - 104.5 %
  • Appearance:
    Passes Test
  • Insoluble Matter:
    ≤0.01 %
  • pH of 5% Solution at 25°C:
    3.0 - 5.0
  • Chloride (Cl):
    ≤0.001 %
  • Phosphate (PO₄):
    ≤0.001 %
  • Calcium (Ca):
    ≤0.005 %
  • Copper (Cu):
    ≤0.005 %
  • Ferric Ion (Fe³⁺):
    ≤0.1 %
  • ACS - Magnesium (Mg):
    ≤0.002 %
  • Manganese (Mn):
    ≤0.05 %
  • Potassium (K):
    ≤0.002 %
  • Sodium (Na):
    ≤0.02 %
  • Zinc (Zn):
    ≤0.005 %

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together