Order Entry
Czech Republic
ContactUsLinkComponent
 
Tetrabutylammonium bromide 50% in aqueous solution, Sigma-Aldrich®
Catalog # SIAL462144-1L
Supplier:  Merck
CAS Number:  
Tetrabutylammonium bromide 50% in aqueous solution, Sigma-Aldrich®
Catalog # SIAL462144-1L
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :322.37 g/mol
Density:1.043 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:216-699-2
CAS Number:1643-19-2
Formula:C₁₆H₃₆BrN
MDL Number:MFCD00011633