Order Entry
Czech Republic
ContactUsLinkComponent
illustra™ MicroSpin™ G-50 columns
illustra™ MicroSpin™ G-50 columns
Supplier:  Cytiva
Certificates
undefined
illustra™ MicroSpin™ G-50 columns
Supplier:  Cytiva

Specifications

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Amplified/modified DNA
  • Sample type:
    PCR and other enzymatic reactions
  • Target molecule:
    DNA
  • Sample size:
    100 µL
  • Description:
    illustra™ MicroSpin™ G-50 columns

Specifications

Related Information

Frequently Bought Together