Caps for bottles, open top, Microlink®, WHEATON®
Dodavatel: DWK Life Sciences
Přejít na:
O tomto produktu
Screw caps are ideal for use with biological media, volatile solvents, corrosive chemicals and sensitive compounds.
- PTFE or PP on the exposed face of the liner imparts an inert, chemical-resistant surface
- Open top caps provide access with a syringe
Ultrasonically welded liner to the cap eliminates the possibility of glue contamination and bond failure on autoclaving. Low-compression-set silicone provides prolonged period of time sample storage over wide temperature ranges.
10 Dostupných variant níže
Product Details & Documents
Screw caps are ideal for use with biological media, volatile solvents, corrosive chemicals and sensitive compounds.
- PTFE or PP on the exposed face of the liner imparts an inert, chemical-resistant surface
- Open top caps provide access with a syringe
Ultrasonically welded liner to the cap eliminates the possibility of glue contamination and bond failure on autoclaving. Low-compression-set silicone provides prolonged period of time sample storage over wide temperature ranges.
Product Family Options
Informace o produktu
- DescriptionThreadColourDostupnostPrice
- Screw cap, solid top, with PP/silicone/PTFE liner20-400WhitePřihlásit se pro dostupnost1 * 48 KS4 000,00 Kč
- Screw cap, solid top, with PP/silicone/PTFE liner20-400WhitePřihlásit se pro dostupnost1 * 600 KS24 870,00 Kč
- Screw cap, solid top, with PP/silicone/PTFE liner24-400WhitePřihlásit se pro dostupnost1 * 48 KS4 760,00 Kč
- Screw cap, open top, with PP/silicone/PTFE liner33-400WhitePřihlásit se pro dostupnost1 * 24 KS4 990,00 Kč
- Screw cap, open top, with PP/silicone/PTFE liner33-400WhitePřihlásit se pro dostupnost1 * 240 KS26 810,00 Kč
- Screw cap, solid top, with PP/silicone/PTFE liner33-430BlackPřihlásit se pro dostupnost1 * 50 KS2 410,00 Kč
- Screw cap, open top, with PP/silicone/PTFE liner33-430BlackPřihlásit se pro dostupnost1 * 50 KS4 070,00 Kč
- Screw cap, open top, with PP/silicone/PTFE liner38-430BlackPřihlásit se pro dostupnost1 * 50 KS4 210,00 Kč
- Screw cap, solid top, with PP/silicone/PTFE liner38-430WhitePřihlásit se pro dostupnost1 * 24 KS3 940,00 Kč
- Screw cap, solid top, with PP/silicone/PTFE liner38-430WhitePřihlásit se pro dostupnost1 * 24 KS5 090,00 Kč