Bottles, narrow neck, round, LDPE, with screw caps, WHEATON®
Supplier: DWK Life Sciences
Certificates
About this item
LDPE, translucent.
- Very flexible with high impact strength
- Excellent for mild and strong buffers, good chemical resistance
- Narrow mouth ideal for liquids
- Pre-attached white polypropylene screw cap with foamed polyethylene liner
Frequently Bought Together