Order Entry
Czech Republic
ContactUsLinkComponent
Dibenzo-21-crown-7-(dibenzo-1,4,7,10,13,16,19-heptaoxacycloheneicosane), Sigma-Aldrich®
Dibenzo-21-crown-7-(dibenzo-1,4,7,10,13,16,19-heptaoxacycloheneicosane), Sigma-Aldrich®
Catalog # SIAL477249-1G
Supplier:  Merck
CAS Number:  
Dibenzo-21-crown-7-(dibenzo-1,4,7,10,13,16,19-heptaoxacycloheneicosane), Sigma-Aldrich®
Catalog # SIAL477249-1G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :404.46 g/mol
 :M-Clarity™ quality level = MQ100
CAS Number:14098-41-0
Formula:C₂₂H₂₈O₇
MDL Number:MFCD00068646