Order Entry
Czech Republic
ContactUsLinkComponent
1,10-Diaza-18-crown-6 (1,4,10,13-tetraoxa-7,16-diazacyclooctadecane kryptofix® 22), Sigma-Aldrich®
1,10-Diaza-18-crown-6 (1,4,10,13-tetraoxa-7,16-diazacyclooctadecane kryptofix® 22), Sigma-Aldrich®
Catalog # SIAL295809-1G
Supplier:  Merck
CAS Number:  
1,10-Diaza-18-crown-6 (1,4,10,13-tetraoxa-7,16-diazacyclooctadecane kryptofix® 22), Sigma-Aldrich®
Catalog # SIAL295809-1G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :262.35 g/mol
 :M-Clarity™ quality level = MQ100
EINECS:245-965-0
CAS Number:23978-55-4
Formula:C₁₂H₂₆N₂O₄
MDL Number:MFCD00005112