
Benzo-18-crown-6 (benzo-1,4,7,10,13,16-hexoxacyclooctadecane) ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:14098-24-9
:
Formula:C₁₆H₂₄O₆
MDL Number:MFCD00062741
:312.36 g/mol
Melting Pt:43…45 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together