Benzo[c]acridine
Catalog # PROOBCR-158
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Typ balení:Glass bottle for solids
- Bal.:100 mg
- Sekvence:2440
Specifications
About this item
:
EINECS:205-930-2
CAS Number:225-51-4
Formula:C₁₇H₁₁N