


Titanium(IV) chloride 1 M in dichloromethane, AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7550-45-0
:
Formula:Cl₄Ti
MDL Number:MFCD00011267
:189.69 g/mol
Storage Temperature:Ambient
EINECS:231-441-9
UN:2922
ADR:6.1,II
:GHS08
:GHS05
:GHS06
Specification Test Results
- Appearance:Light yellow to dark brown Solution
- Titration Argentometric:0.95 to 1.10 M
Specification Test Results
Related Information
Frequently Bought Together