

tert-Butyl carbazate 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:870-46-2
:
Formula:C₅H₁₂N₂O₂
MDL Number:MFCD00007593
:132.16 g/mol
Boiling Pt:63…65 °C (.1 mmHg)
Melting Pt:37…41 °C
Density:1.02 g/cm³
Flash Pt:91 °C
Storage Temperature:Ambient
EINECS:212-795-3
UN:2926
ADR:4.1,II
:GHS06
:GHS02
Specification Test Results
- Appearance:White to light yellow Lumps
- Infrared spectrum:Conforms
- Melting point:37 to 41 °C
- Titration with HClO4:≥98.5 %
Specification Test Results
Frequently Bought Together