Order Entry
Czech Republic
ContactUsLinkComponent
Di-tert-butyl pyrocarbonate 2 mol/l in ethyl acetate, Sigma-Aldrich®
Di-tert-butyl pyrocarbonate 2 mol/l in ethyl acetate, Sigma-Aldrich®
Catalog # SIAL703753-100ML
Supplier:  Merck
CAS Number:  
undefined
Di-tert-butyl pyrocarbonate 2 mol/l in ethyl acetate, Sigma-Aldrich®
Catalog # SIAL703753-100ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :218.25 g/mol
 :M-Clarity™ quality level = MQ100
EINECS:246-240-1
CAS Number:24424-99-5
Formula:C₁₀H₁₈O₅
MDL Number:MFCD00008805