2,2-Difluoropropionic acid, Sigma-Aldrich®
Catalog # SIAL684686-250MG
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:110.06 g/mol
ADR:8,II
:M-Clarity™ quality level = MQ100
UN:3261
CAS Number:373-96-6
Formula:C₃H₄F₂O₂
MDL Number:MFCD03093764