Lithium azide 20% in aqueous solution, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:19597-69-4
:
Formula:LiN₃
MDL Number:MFCD00041956
:48.96 g/mol
:M-Clarity™ quality level = MQ200
EINECS:243-177-1
Frequently Bought Together