
3,3'-Dithiodipropionic acid 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1119-62-6
:
Formula:C₆H₁₀O₄S₂
MDL Number:MFCD00002780
Melting Pt:155…158 °C
Storage Temperature:Ambient
EINECS:214-284-0
UN:3335
ADR:9,III
:210.28 g/mol
:GHS07
Specification Test Results
- Appearance:White Powder
- Infrared spectrum:Conforms
- Melting point:154 to 158 °C
- Titration with NaOH:≥97.5 %
Specification Test Results
Frequently Bought Together