
2-Isobutylpyridine ≥99%
Catalog # B22349.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
CAS Number:6304-24-1
:
Formula:C₉H₁₃N
Density:0.899
:135.21 g/mol
EINECS:228-606-2
:GHS07
Storage Temperature:Ambient
MDL Number:MFCD02262142