Order Entry
Czech Republic
ContactUsLinkComponent
2,6-Dimethoxypyridine ≥98%
2,6-Dimethoxypyridine ≥98%
Catalog # B22746.22
CAS Number:  
2,6-Dimethoxypyridine ≥98%
Catalog # B22746.22
CAS Number:  

Specifications

  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

CAS Number:6231-18-1
 :
Formula:C₇H₉NO₂
MDL Number:MFCD00006266
 :139.15 g/mol
Boiling Pt:178 °C (1013 hPa)
Density:1.05 g/cm³ (20 °C)
Flash Pt:61 °C (141 °F)
EINECS:228-334-4