
2,6-Dichloro-3-nitropyridine ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:16013-85-7
:
Formula:C₅H₂Cl₂N₂O₂
MDL Number:MFCD00006234
:192.99 g/mol
Melting Pt:62…65 °C
Flash Pt:>110 °C (230 °F)
EINECS:240-151-1
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together