
2,6-Dichloro-3-nitropyridine 90% tech.
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:16013-85-7
:
Formula:C₅H₂Cl₂N₂O₂
MDL Number:MFCD00006234
:192.99 g/mol
Melting Pt:58…68 °C
Flash Pt:>110 °C
EINECS:240-151-1
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Yellow to orange-brown Powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Melting point:58 to 68 °C
- Water:≤10 % (K.F.)
- GC:≥88 %
Specification Test Results
Frequently Bought Together