2,6-Dibromopyridine ≥98%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 98% CAS No.: 626-05-1 Molecular Formula: C5H3Br2N Formula Weight: 236.90 Flash Point: 213°(415°F) Odour: Recognizable Boiling Point: 255° Beilstein Reference: 108922 UN No.: 2811 MDL No.: MFCD00006223
- CAS Number:626-05-1
- Formula:C₅H₃Br₂N
- MDL Number:MFCD00006223
- Molecular Weight:236.89 g/mol
- Boiling Pt:255 °C
- Melting Pt:117…120 °C
- Flash Pt:213 °C (415 °F)
- Storage Temperature:Ambient
- EINECS:210-926-9
- UN:2811
- ADR:6.1,II
3 Options Available Below
Product Details & Documents
Purity: 98% CAS No.: 626-05-1 Molecular Formula: C5H3Br2N Formula Weight: 236.90 Flash Point: 213°(415°F) Odour: Recognizable Boiling Point: 255° Beilstein Reference: 108922 UN No.: 2811 MDL No.: MFCD00006223
- CAS Number:626-05-1
- Formula:C₅H₃Br₂N
- MDL Number:MFCD00006223
- Molecular Weight:236.89 g/mol
- Boiling Pt:255 °C
- Melting Pt:117…120 °C
- Flash Pt:213 °C (415 °F)
- Storage Temperature:Ambient
- EINECS:210-926-9
- UN:2811
- ADR:6.1,II



