Order Entry
Czech Republic
ContactUsLinkComponent
Tetrahydrofuran, EMPARTA® ACS analytical reagent, Supelco®
Flammable
Health
Irritant
Tetrahydrofuran, EMPARTA® ACS analytical reagent, Supelco®
Catalog # 1.07025.2500
Supplier:  Merck
CAS Number:  
undefined
Flammable
Health
Irritant
Tetrahydrofuran, EMPARTA® ACS analytical reagent, Supelco®
Catalog # 1.07025.2500
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    -108.5 °C
  • Molar Mass:
    72.11 g/mol
  • Boiling Point:
    65 - 66 °C (1013 hPa)
  • Vapor Pressure:
    173 hPa (20 °C)
  • Flash Point:
    -21.5 °C
  • Refractive Index:
    1.407 (20 °C)
  • Explosion Limit:
    1.5 - 12.4 %(V)
  • Density:
    0.89 g/cm3 (20 °C)
  • pH:
    7 - 8 (200 g/l, H2O, 20 °C)
  • Ignition Point:
    215 °C
  • Evaporation Number:
    2.3

Specification Test Results

About this item

 :
 :72.11 g/mol
Merck Index:12,09351
Boiling Pt:66 °C (1013 hPa)
Melting Pt:–108.5 °C
Density:0.888 g/cm³ (20 °C)
Flash Pt:–21.5 °C
Storage Temperature:Ambient
EINECS:203-726-8
UN:2056
ADR:3,II
 :M-Clarity™ quality level = MQ200
REACH:05-2114615188-48
 :GHS02
 :GHS08
 :GHS07
CAS Number:109-99-9
Formula:C₄H₈O
MDL Number:MFCD00005356