Tetrahydrofuran ≥99.7% unstabilised for spectrophotometry
:109-99-9
:
:C₄H₈O
:MFCD00005356
:72.11 g/mol
:14,9211
:66 °C
:–108 °C
:0.889
:–17 °C (1 °F)
:Ambient
:203-726-8
:2056
:3,II
:05-2114615188-48
:GHS02
:GHS08