



Naphthalene for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:91-20-3
:
Formula:C₁₀H₈
MDL Number:MFCD00001742
:128.17 g/mol
Merck Index:12,06457
Boiling Pt:218 °C (1013 hPa)
Melting Pt:79…82 °C
Density:1.145 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:202-049-5
UN:1334
ADR:4.1,III
:M-Clarity™ quality level = MQ200
Flash Pt:79 °C
:GHS02
:GHS08
:GHS07
:GHS09
Specification Test Results
- Solubility:0.03 g/l (25 °C)
- Melting Point:79 - 82 °C
- Molar Mass:128.17 g/mol
- Bulk Density:550 - 600 mg/m3
- Boiling Point:218 °C
- Vapor Pressure:0.08 hPa (20 °C)
- Refractive Index:80 °C
- Explosion Limit:0.9 - 5.9 %(V)
- Density:1.15 g/cm3 (20 °C)
- Ignition Point:540 °C
- Hazard Class:4.1
- Hazard Class Description:Flammable Solid
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC8.20846.0100_TDS_EN ProductPage_820846
Product specification
msds_820846
SDS
Frequently Bought Together