

2-Naphthol
Catalog # EHERC15430500
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,5 g
- Seq:3035
Specifications
About this item
:
Merck Index:12,06471
Boiling Pt:286 °C (1013 hPa)
Melting Pt:121.6 °C
Density:1.217 g/cm³ (20 °C)
Flash Pt:153 °C
Storage Temperature:Ambient
EINECS:205-182-7
UN:3077
ADR:9,III
:144.17 g/mol
:GHS07
:GHS09
CAS Number:135-19-3
Formula:C₁₀H₈O
MDL Number:MFCD00004067