

1,5-Dinitronaphthalene ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:605-71-0
:
Formula:C₁₀H₆N₂O₄
MDL Number:MFCD00003916
Boiling Pt:390 °C (1013 hPa)
Melting Pt:214…218 °C
Density:1.586
:218.17 g/mol
Storage Temperature:Ambient
EINECS:210-095-2
:GHS08
:GHS07
Related Information
Frequently Bought Together