2,6-Naphthalenedicarboxylic acid ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1141-38-4
:
Formula:C₁₂H₈O₄
MDL Number:MFCD00004105
:216.19 g/mol
Boiling Pt:422 °C (1013 hPa)
Melting Pt:>300 °C
Storage Temperature:Ambient
EINECS:214-527-0
Related Information
Frequently Bought Together