Order Entry
Czech Republic
ContactUsLinkComponent
 
 
1,4-Dibromonaphthalene 98+%
 
undefined
 
1,4-Dibromonaphthalene 98+%

 

  • Appearance:
    White to light beige Crystalline powder or crystals
  • Infrared spectrum:
    Conforms
  • Melting point:
    79 to 83 °C
  • GC:
    ≥98.0 %

 

 

Frequently Bought Together