
Zinc(II) telluride ≥99.99% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1315-11-3
:
Formula:TeZn
MDL Number:MFCD00043079
:192.99 g/mol
Merck Index:14,10162
Melting Pt:1240 °C
Density:5.9
EINECS:215-260-2
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together