
Zinc oxide ≥99.9% (metals basis), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 6.35 mm (0.250 in)
Catalog # 41124.KS
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1
- Seq:0001
Specifications
About this item
:
:81.39 g/mol
Merck Index:13,10200
Melting Pt:1975 °C
Density:5.61 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:215-222-5
UN:3077
ADR:9,III
REACH:01-2119463881-32
:GHS09
CAS Number:1314-13-2
Formula:ZnO
MDL Number:MFCD00011300