
Praseodymium(III,IV) oxide 99.9% (trace metals basis)
Catalog # ACRO194620500
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g
- Seq:3368
Specifications
Specification Test Results
- Appearance:Dark green to dark brown to black Powder
- Total conc. of imp.as REO:≤0.1 %
Specification Test Results
About this item
:
:1021.44 g/mol
Melting Pt:2012…2072 °C
EINECS:234-857-9
:GHS07
Storage Temperature:Ambient
CAS Number:12037-29-5
Formula:O₁₁Pr₆
MDL Number:MFCD00011178