1,3-Indandione for synthesis, Sigma-Aldrich®
Catalog # 8.20698.0010
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 g
- Seq:3236
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:(20 °C) insoluble
- Melting Point:129 - 132 °C
- Molar Mass:146.14 g/mol
- Synonyms:1,3-Hydrindenedione, 1,3-Dioxoindane
- Hill Formula:C9H6O2
Specification Test Results
About this item
Synonyms: 1,3-Indanedione, Indan-1,3-dione, 1H-Indene-1,3(2H)-dione
:
:146.15 g/mol
Boiling Pt:283 °C (1013 hPa)
Melting Pt:129…132 °C
Density:1.37 g/cm³ (21 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:210-109-7
CAS Number:606-23-5
Formula:C₉H₆O₂
MDL Number:MFCD00003779