


Selenium chloride ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10025-68-0
:
Formula:SeCl
MDL Number:MFCD00011226
:228.83 g/mol
Merck Index:14,8432
Boiling Pt:127 °C (dec.)
Melting Pt:–85 °C
Density:2.77
Storage Temperature:Ambient
UN:2922
ADR:8,II
:ampouled under argon
EINECS:233-037-8
:GHS06
:GHS05
:GHS08
Related Information
Frequently Bought Together