


Phosphorus pentasulphide for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:1314-80-3
:
Formula:P₂S₅
MDL Number:MFCD00011441
:222.28 g/mol
Merck Index:12,07511
Boiling Pt:513 °C (1013 hPa)
Melting Pt:284…287 °C
Density:2.08 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1340
ADR:4.3 (4.1),II
:M-Clarity™ quality level = MQ200
EINECS:215-242-4
:GHS02
:GHS07
:GHS09
Specification Test Results
- Solubility:(20 °C) (decomposition)
- Melting Point:284 - 287 °C
- Molar Mass:222.28 g/mol
- Bulk Density:900 - 1100 kg/m3
- Boiling Point:513 - 515 °C (1013 hPa)
- Density:2.08 g/cm3 (20 °C)
- pH:1 (10 g/l, H2O, 20 °C)
- Ignition Point:142 °C
- Hazard Class:4.3
- Hazard Class Description:Dangerous When Wet Material
Specification Test Results
Frequently Bought Together