Order Entry
Czech Republic
ContactUsLinkComponent
 
Toxic
Corrosive
Health
Phosphorus(V) chloride ≥98%
Catalog # 10523.22
CAS Number:  
Toxic
Corrosive
Health
Phosphorus(V) chloride ≥98%
Catalog # 10523.22
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

CAS Number:10026-13-8
 :
Formula:Cl₅P
MDL Number:MFCD00011439
 :208.24 g/mol
Merck Index:14,7351
Melting Pt:162 °C (subl.)
Density:2.114
EINECS:233-060-3
UN:1806
ADR:8,II
 :GHS06
 :GHS05
 :GHS08
Storage Temperature:Ambient