Silica gel, Pore size: 60 Å 230-400 mesh, high purity for flash chromatography, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:112926-00-8
:7631-86-9
Formula:SiO₂
MDL Number:MFCD00011232
:60.08 g/mol
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
Frequently Bought Together