Order Entry
Czech Republic
ContactUsLinkComponent
Silica gel, Pore size: 60 Å 0.063-0.200 mm (70-230 mesh ASTM) for column chromatography, Millipore®
Silica gel, Pore size: 60 Å 0.063-0.200 mm (70-230 mesh ASTM) for column chromatography, Millipore®
  1.07734.2500
 :  
Silica gel, Pore size: 60 Å 0.063-0.200 mm (70-230 mesh ASTM) for column chromatography, Millipore®
  1.07734.2500
 :  

 

  • Pack type:
    Plastic bottle for solids
  • Pk:
    2,5 kg
  • Seq:
    4076

 

 

  • pH-value (10% suspension):
    6.5 - 7.5
  • Fe (Iron):
    ≤0.02%
  • Pore volume (N₂-isotherm):
    0.74 - 0.84 ml/g
  • Specific surface area (according to BET; 5-Pt. measurement):
    480 - 540 m²/g
  • Loss on drying (150 °C):
    ≤7.0%
  • Particle Size (d10):
    75 - 95 µm
  • Particle Size (d50):
    125 - 150 µm
  • Particle Size (d90):
    215 - 245 µm

 

 

 :
 :60.08 g/mol
 :Ambient
 :231-545-4
 :63231-67-4
 :112926-00-8
 :SiO₂
 :MFCD00011232