Order Entry
Czech Republic
ContactUsLinkComponent
Silica gel, Pore size: 60 Å 40-60 µm, Ultrapure for column chromatography
Silica gel, Pore size: 60 Å 40-60 µm, Ultrapure for column chromatography
Certificates
undefined
Silica gel, Pore size: 60 Å 40-60 µm, Ultrapure for column chromatography

Specification Test Results

  • Appearance:
    White Powder
  • Particle size:
    ≤10 % (Undersize)
  • Specific surface area:
    460 to 510 m²/g
  • Pore volume:
    0.75 to 0.80 ml/g
  • pH:
    6.5 to 7.0 (10 % suspension in water)
  • Volatile matter:
    ≤6.0 % (160°C)

Specification Test Results

Related Information

Frequently Bought Together